Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxacillin sodium monohydrate | 7240-38-2 | 99.6% | 1 ML
SDP

Supplier:  Medchemexpress LLC HYB046510MM/1ML

Encompass_Preferred

Oxacillin sodium monohydrate is an antibiotic used in the study of resistant staphylococci infections, similar to Flucloxacillin.

  • Purity of 99.62%
  • Molecular weight of 441.43
  • Chemical formula C19H20N3NaO6S
  • Appearance as a white to off-white solid
  • Soluble in H2O (≥ 100 mg/mL) and DMSO (50 mg/mL)
  • Storage at 4°C for solid, and -80°C (6 months) or -20°C (1 month) in solvent under sealed, moisture-free conditions
  • Can be prepared using various in vivo dissolution protocols

Catalog No. 50-004-08422


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.