Learn More
Medchemexpress LLC Oxacillin sodium salt | 1173-88-2 | 99.5% | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HYB092510MM/1ML
Encompass_Preferred
Oxacillin sodium salt is an orally active synthetic penicillin that demonstrates good bactericidal activity against staphylococci and other gram-positive pathogens. It is intended for research use only.
  • Orally active synthetic penicillin
  • Good bactericidal activity against staphylococci
  • Effective against gram-positive pathogens
Catalog No. 50-004-07694