Learn More
Alkali Scientific Oxacillin Sodium Salt Monohydrate, 1 G
SDP
Catalog No. 5000660976
Encompass_Preferred
Oxacillin Sodium Salt Monohydrate, 1G, is a laboratory-grade antibiotic used primarily for antimicrobial testing and research applications. As a penicillin-class antibiotic, it is effective against a variety of Gram-positive bacteria, particularly Staphylococcus species. Oxacillin works by inhibiting bacterial cell wall synthesis, which leads to cell death. This product is widely used in laboratory settings for studying bacterial resistance, particularly for assessing methicillin resistance in staphylococci. The 1g quantity is ideal for smaller scale studies and testing applications where precise antibiotic measurement is needed for microbial assays, including disk diffusion and dilution testing methods.
Catalog No. 50-006-60976 Supplier Alkali Scientific Supplier No. OXA001CP
Oxacillin Sodium Salt Monohydrate, 1G, is a laboratory-grade antibiotic used primarily for antimicrobial testing and research applications. As a penicillin-class antibiotic, it is effective against a variety of Gram-positive bacteria, particularly Staphylococcus species. Oxacillin works by inhibiting bacterial cell wall synthesis, which leads to cell death. This product is widely used in laboratory settings for studying bacterial resistance, particularly for assessing methicillin resistance in staphylococci. The 1g quantity is ideal for smaller scale studies and testing applications where precise antibiotic measurement is needed for microbial assays, including disk diffusion and dilution testing methods.