Learn More
Medchemexpress LLC Oxadiazon (Standard) | 19666-30-9 | 98.6% | 250 MG
SDP
Catalog No. 5000413956
Encompass_Preferred
Oxadiazon (Standard) is the analytical standard for Oxadiazon and is intended for research and analytical applications. It functions as an orally active herbicide that controls weeds and has been shown to reduce hepatic cytochrome P450 levels, increase palmitoyl CoA oxidase, acetyl carnitine transferase, and hepatic ALAS activity. It also induces hepatomegaly and experimental porphyria, and may be used in neurodegenerative disease research.
  • Serves as a reference standard for qualitative, quantitative, and methodological research experiments.
  • Commonly used in techniques such as HPLC, GC, and MS.
  • Functions as an orally active herbicide for weed control.
  • Reduces hepatic cytochrome P450 levels and increases palmitoyl CoA oxidase, acetyl carnitine transferase, and hepatic ALAS activity.
  • May be utilized in studies related to neurodegenerative diseases.
Catalog No. 50-004-13956 Supplier Medchemexpress LLC Supplier No. 250MGHYB1880R
Oxadiazon (Standard) is the analytical standard for Oxadiazon and is intended for research and analytical applications. It functions as an orally active herbicide that controls weeds and has been shown to reduce hepatic cytochrome P450 levels, increase palmitoyl CoA oxidase, acetyl carnitine transferase, and hepatic ALAS activity. It also induces hepatomegaly and experimental porphyria, and may be used in neurodegenerative disease research.
  • Serves as a reference standard for qualitative, quantitative, and methodological research experiments.
  • Commonly used in techniques such as HPLC, GC, and MS.
  • Functions as an orally active herbicide for weed control.
  • Reduces hepatic cytochrome P450 levels and increases palmitoyl CoA oxidase, acetyl carnitine transferase, and hepatic ALAS activity.
  • May be utilized in studies related to neurodegenerative diseases.