Learn More
Selleck Chemical LLC Oxalacetic acid
SDP
Catalog No. 5000000786
Encompass_Preferred
Oxalacetic acid (Oxaloacetic acid 2-Oxosuccinic acid Ketosuccinic acid) is an intermediate of the citric acid cycle where it reacts with acetyl-CoA to form citrate catalysed by citrate synthase It is also involved in gluconeogenesis urea cycle glyoxylate cycle amino acid synthesis and fatty acid synthesis Oxaloacetate is also a potent inhibitor of Complex II
Catalog No. 50-000-00786 Supplier Selleck Chemical LLC Supplier No. S61121G
Oxalacetic acid (Oxaloacetic acid 2-Oxosuccinic acid Ketosuccinic acid) is an intermediate of the citric acid cycle where it reacts with acetyl-CoA to form citrate catalysed by citrate synthase It is also involved in gluconeogenesis urea cycle glyoxylate cycle amino acid synthesis and fatty acid synthesis Oxaloacetate is also a potent inhibitor of Complex II