Learn More
Selleck Chemical LLC Oxalacetic acid
SDP
Supplier:  Selleck Chemical LLC S611225MG
Encompass_Preferred
Oxalacetic acid (Oxaloacetic acid 2-Oxosuccinic acid Ketosuccinic acid) is an intermediate of the citric acid cycle where it reacts with acetyl-CoA to form citrate catalysed by citrate synthase It is also involved in gluconeogenesis urea cycle glyoxylate cycle amino acid synthesis and fatty acid synthesis Oxaloacetate is also a potent inhibitor of Complex II
Catalog No. 50-000-04239