Learn More
Sigma Aldrich Fine Chemicals Biosciences Oxalate Assay Kit sufficie
Catalog No. 502904602
Encompass
Oxalate or Oxalic Acid is a metabolic breakdown product of the Krebs Cycle in eukaryotes and the glyoxylate cycle in other microorganisms. It can be found in the urine of humans and other mammals. Oxalate concentration can be used as a measure of kidney function where a high level of oxalate is an indicator for kidney stones which are primarily made of the insoluble salt calcium oxalate. Measuring oxalate is more accurate than measuring calcium as a marker for kidney stones because calcium is excreted at high concentrations even in normal urine.
Catalog No. 50-290-4602 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MAK3151KT
Oxalate or Oxalic Acid is a metabolic breakdown product of the Krebs Cycle in eukaryotes and the glyoxylate cycle in other microorganisms. It can be found in the urine of humans and other mammals. Oxalate concentration can be used as a measure of kidney function where a high level of oxalate is an indicator for kidney stones which are primarily made of the insoluble salt calcium oxalate. Measuring oxalate is more accurate than measuring calcium as a marker for kidney stones because calcium is excreted at high concentrations even in normal urine.