Learn More
Electron Microscopy Sciences Oxalic Acid 5% Aqueous 500 ML
SDP
Catalog No. 5031877
Encompass_Preferred
Used in Ptah Method for Central Nervous Tissue for staining microglia, fibroglia, fibrin and coarse fibrils. References: AFIP Manual of Histologic Staining Techniques; (ed) G. Luna, 3rd Ed. McGraw-Hill Publications, c. 1968, p. 199
Catalog No. 50-318-77 Supplier Electron Microscopy Sciences Supplier No. 2665803
Used in Ptah Method for Central Nervous Tissue for staining microglia, fibroglia, fibrin and coarse fibrils. References: AFIP Manual of Histologic Staining Techniques; (ed) G. Luna, 3rd Ed. McGraw-Hill Publications, c. 1968, p. 199