Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid ACS AR Crystal, Macron Fine Chemicals™

Catalog No. MK2752500 Shop All Avantor Products
Change view
Click to view available options
POLYSTORMOR; 12 x 500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
MK-2752-500 POLYSTORMOR; 12 x 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-2752-500 Supplier Avantor Supplier No. 275204

Specifications

CAS 6153-56-6
Molecular Formula C2H6O6
Synonym oxalic acid dihydrate, ethanedioic acid, dihydrate, unii-0k2l2ij59o, ethanedioic acid dihydrate, oxalic acid, dihydrate, oxalicacid dihydrate, dihydrate oxalic acid, oxalic acid di-hydrate, pubchem17416, acmc-209mtm
InChI Key GEVPUGOOGXGPIO-UHFFFAOYSA-N
SMILES C(=O)(C(=O)O)O.O.O
IUPAC Name oxalic acid;dihydrate
Molecular Weight (g/mol) 126.064
PubChem CID 61373
Formula Weight 126.07
Grade ACS
Quantity 12 x 500 g
Packaging POLYSTORMOR
Physical Form Crystalline
Chemical Name or Material Oxalic Acid, Dihydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.