Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxalic acid ammonium | 1113-38-8 | >99.0% | 124.10 g/mol | C2H8N2O4 | 500 G
SDP

Catalog No. 5000209147
Encompass_Preferred

Oxalic acid ammonium is the ammonium salt of oxalic acid supplied as a high-purity research reagent. It is used in laboratory and plant-science studies for its acidity, reducing, and chelating properties.

  • Ammonium salt of oxalic acid, suitable for research use.
  • Purity >99.0%.
  • Molecular weight 124.10 g/mol; formula C2H8N2O4.
  • Available in a 500 G package.
  • Useful for studies of plant physiology, metal chelation, and analytical applications.

Catalog No. 50-002-09147 Supplier Medchemexpress LLC Supplier No. HYW041831500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxalic acid ammonium is the ammonium salt of oxalic acid supplied as a high-purity research reagent. It is used in laboratory and plant-science studies for its acidity, reducing, and chelating properties.

  • Ammonium salt of oxalic acid, suitable for research use.
  • Purity >99.0%.
  • Molecular weight 124.10 g/mol; formula C2H8N2O4.
  • Available in a 500 G package.
  • Useful for studies of plant physiology, metal chelation, and analytical applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.