Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxalic acid ammonium | 1113-38-8 | MFCD00013308 | ≥99.0% | 124.10 g/mol | C2H8N2O4 | 250 G
SDP

Catalog No. 5000208882
Encompass_Preferred

Oxalic acid ammonium is the ammonium salt of oxalic acid used as a reagent for research and analytical applications. It has high acidity, reducibility, and chelation ability, and is commonly used in studies of plant growth and stress responses.

  • Purity ≥99.0%
  • CAS number 1113-38-8
  • Molecular formula C2H8N2O4; molecular weight 124.10 g/mol
  • Available pack sizes: 250 g, 500 g, 1 kg
  • Functions as a chelating agent and reagent in biochemical and plant studies

Catalog No. 50-002-08882 Supplier Medchemexpress LLC Supplier No. 250GHYW041831
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxalic acid ammonium is the ammonium salt of oxalic acid used as a reagent for research and analytical applications. It has high acidity, reducibility, and chelation ability, and is commonly used in studies of plant growth and stress responses.

  • Purity ≥99.0%
  • CAS number 1113-38-8
  • Molecular formula C2H8N2O4; molecular weight 124.10 g/mol
  • Available pack sizes: 250 g, 500 g, 1 kg
  • Functions as a chelating agent and reagent in biochemical and plant studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.