Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid Concentrate, 0.005M, Solstice

Catalog No. 6002682 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Pc.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-026-82 1 Pc.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-026-82 Supplier Solstice Supplier No. 382551EA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for 1L standard solution, 0.005 M (COOH)2 (0.01N)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-62-7
Molecular Formula C2H2O4
Molecular Weight (g/mol) 90.034
MDL Number MFCD00002573
InChI Key MUBZPKHOEPUJKR-UHFFFAOYSA-N
Synonym ethanedioic acid, aktisal, aquisal, oxiric acid, oxalate, oxalsaeure, oxaalzuur, kyselina stavelova, acide oxalique, acido ossalico
PubChem CID 971
ChEBI CHEBI:16995
IUPAC Name oxalic acid
SMILES C(=O)(C(=O)O)O

Specifications

Quantity 1 Pc.
Linear Formula HOOCCOOH
UN Number NONH for all modes of transport
Formula Weight 90.03g/mol
Chemical Name or Material Oxalic acid concentrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.