Learn More
Medchemexpress LLC Oxalic acid diammonium monohydrate | 6009-70-7 | MFCD00149694 | 98.0% | 142.11 g/mol | C2H10N2O5 | 1g
SDP
Catalog No. 5000266989
Encompass_Preferred
Oxalic Acid is a strong dicarboxylic acid occurring in many plants and vegetables and can be used as an analytical reagent and general reducing agent
Catalog No. 50-002-66989 Supplier Medchemexpress LLC Supplier No. HYY0262A1G
Oxalic Acid is a strong dicarboxylic acid occurring in many plants and vegetables and can be used as an analytical reagent and general reducing agent