Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oxalic Acid Dihydrate

Catalog No. O0376100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 g
1 kg
3 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O0376100G 1 kg
O03761KG 3 kg
O03763KG 500 g
O0376500G 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. O0376100G Supplier Sigma Aldrich Supplier No. O0376100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Oxalic Acid Dihydrate

Specifications

Melting Point 104°C to 106°C
CAS 6153-56-6
Molecular Formula C2H2O4 -+ 2H2O
Linear Formula HO2CCO2H -+ 2H2O
MDL Number MFCD00149102
Synonym Ethanedioic acid dihydrate
Molecular Weight (g/mol) 126.07 g/mol
Quantity 1 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.