Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxalic Acid Dihydrate, Reagents

Catalog No. 01337577
Change view
Click to view available options
Quantity:
100 g
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-577 500 g
01-337-580 100 g
01-337-578 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-577 Supplier Reagents Holdings Llc Supplier No. C2110500500A5
Add to Cart
Add to Cart

Oxalic Acid Dihydrate, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Oxalic Acid Dihydrate
CAS 6153-56-6
Melting Point 101°C to 102°C
Color Colorless/White
Molecular Formula C2H6O6, 2H2O
Synonym Ethanedioic Acid dihydrate
Molecular Weight (g/mol) 162.1
Grade Pure
Quantity 500 g
Packaging Poly Bottle
Physical Form Solid
Specific Gravity 1.653
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.