Learn More
Medchemexpress LLC HY-17371 100mg , Oxaliplatin CAS:61825-94-3 Purity:>98%
SDP
Catalog No. 502259153
Encompass_Preferred
HY-17371 100mg Oxaliplatin CAS: 61825-94-3 Oxaliplatin is a DNA synthesis inhibitor. Oxaliplatin causes DNA crosslinking damage, prevents DNA replication and transcription and induces apoptosis. Oxaliplatin can be used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-225-9153 Supplier Medchemexpress LLC Supplier No. HY17371100MG
HY-17371 100mg Oxaliplatin CAS: 61825-94-3 Oxaliplatin is a DNA synthesis inhibitor. Oxaliplatin causes DNA crosslinking damage, prevents DNA replication and transcription and induces apoptosis. Oxaliplatin can be used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.