Learn More
TARGETMOL CHEMICALS INC Oxaliplatin 200MG
SDP
Catalog No. 502880682
Encompass_Preferred
Also available in 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxaliplatin (L-OHP) is a DNA alkylating agent, an inhibitor of DNA synthesis. Oxaliplatin causes DNA cross-linking damage, preventing DNA replication and transcription and leading to cell death. Oxaliplatin induces autophagy. Purity 99.93%
Catalog No. 50-288-0682 Supplier TARGETMOL CHEMICALS INC Supplier No. T0164200MG
Also available in 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Oxaliplatin (L-OHP) is a DNA alkylating agent, an inhibitor of DNA synthesis. Oxaliplatin causes DNA cross-linking damage, preventing DNA replication and transcription and leading to cell death. Oxaliplatin induces autophagy. Purity 99.93%