Learn More
Medchemexpress LLC HY-17371 500mg , Oxaliplatin CAS:61825-94-3 Purity:98%
SDP
Catalog No. 501492332
Encompass_Preferred
Medchemexpress, HY-17371 500mg Oxaliplatin CAS:61825-94-3 Formula:C8H14N2O4Pt Purity:98% Oxaliplatin inhibits DNA synthesis by conforming DNA adducts. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2332 Supplier Medchemexpress LLC Supplier No. HY17371500MG
Medchemexpress, HY-17371 500mg Oxaliplatin CAS:61825-94-3 Formula:C8H14N2O4Pt Purity:98% Oxaliplatin inhibits DNA synthesis by conforming DNA adducts. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.