Learn More
Enzo Life Sciences Oxaliplatin (5mg). CAS: 61825-94-3
SDP
Supplier:  Enzo Life Sciences ALX400042M005
Encompass_Preferred
Platinum analog which has wide spectrum anticancer activity. It is found to be more active against human melanoma cell lines, and has better biochemical, pharmacological and cytotoxic properties than cisplatin and carboplatin. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline powder. Solubility: Slightly soluble in water (7.9mg/ml); very slightly soluble in methanol; almost insoluble in 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9083