Learn More
Medchemexpress LLC HY-17371 5mg , Oxaliplatin CAS:61825-94-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY173715MG
Encompass_Preferred
HY-17371 5mg Oxaliplatin CAS: 61825-94-3 Oxaliplatin is a DNA synthesis inhibitor. Oxaliplatin causes DNA crosslinking damage, prevents DNA replication and transcription and induces apoptosis. Oxaliplatin can be used for cancer research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9027