Learn More
Selleck Chemical LLC Oxaliplatin 200mg 61825-94-3 L-OHP
SDP
Catalog No. 507471
Encompass_Preferred
Oxaliplatin (L-OHP) is a DNA alkylating agent that activates autophagy. Oxaliplatin inhibits DNA synthesis by conforming DNA adducts in RT4, TCCSUP, A2780, HT-29, U-373MG, U-87MG, SK-MEL-2, and HT-144 cells. DMF is recommended for dissolution. Solutions are best fresh-prepared. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-747-1 Supplier Selleck Chemical LLC Supplier No. S1224200MG
Oxaliplatin (L-OHP) is a DNA alkylating agent that activates autophagy. Oxaliplatin inhibits DNA synthesis by conforming DNA adducts in RT4, TCCSUP, A2780, HT-29, U-373MG, U-87MG, SK-MEL-2, and HT-144 cells. DMF is recommended for dissolution. Solutions are best fresh-prepared. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.