Learn More
Medchemexpress LLC Oxaloacetic acid (2-Oxosuccinic acid) | 328-42-7 | 99.9% | 132.07 | 10 MG
SDP
Catalog No. 5000367067
Encompass_Preferred
Oxaloacetic acid (2-Oxosuccinic acid) is a metabolic intermediate that participates in various biochemical pathways, including the citric acid cycle, gluconeogenesis, the urea cycle, the glyoxylate cycle, amino acid synthesis, and fatty acid synthesis. This compound plays a role in facilitating the clearance of reactive oxygen species (ROS) and enhancing mitochondrial function.
  • Metabolic intermediate
  • Involved in citric acid cycle, gluconeogenesis, urea cycle, glyoxylate cycle, amino acid synthesis, and fatty acid synthesis
  • Facilitates the clearance of reactive oxygen species (ROS)
  • Improves mitochondrial function
  • For research use only
  • Appears as a white to off-white solid
  • Endogenous metabolite
Catalog No. 50-003-67067 Supplier Medchemexpress LLC Supplier No. HYW010382R10MG
Oxaloacetic acid (2-Oxosuccinic acid) is a metabolic intermediate that participates in various biochemical pathways, including the citric acid cycle, gluconeogenesis, the urea cycle, the glyoxylate cycle, amino acid synthesis, and fatty acid synthesis. This compound plays a role in facilitating the clearance of reactive oxygen species (ROS) and enhancing mitochondrial function.
  • Metabolic intermediate
  • Involved in citric acid cycle, gluconeogenesis, urea cycle, glyoxylate cycle, amino acid synthesis, and fatty acid synthesis
  • Facilitates the clearance of reactive oxygen species (ROS)
  • Improves mitochondrial function
  • For research use only
  • Appears as a white to off-white solid
  • Endogenous metabolite