Learn More
Medchemexpress LLC Oxaloacetic acid | 328-42-7 | 132.07 | 1 ML
SDP
Catalog No. 5000367041
Encompass_Preferred
Oxaloacetic acid (2-Oxosuccinic acid) is a metabolic intermediate involved in various cycles such as the citric acid cycle, gluconeogenesis, the urea cycle, the glyoxylate cycle, amino acid synthesis, and fatty acid synthesis. It facilitates the clearance of reactive oxygen species (ROS) and improves mitochondrial function.
  • For research use only
  • Alleviates Paraquat-induced acute lung injury
  • Improves survival rate in mice
  • Involved in metabolic enzyme/protease pathway
  • Involved in immunology/inflammation pathway
  • Involved in NF-κB pathway
Catalog No. 50-003-67041 Supplier Medchemexpress LLC Supplier No. HYW01038210MM/1ML
Add to Cart
Add to Cart
Oxaloacetic acid (2-Oxosuccinic acid) is a metabolic intermediate involved in various cycles such as the citric acid cycle, gluconeogenesis, the urea cycle, the glyoxylate cycle, amino acid synthesis, and fatty acid synthesis. It facilitates the clearance of reactive oxygen species (ROS) and improves mitochondrial function.
  • For research use only
  • Alleviates Paraquat-induced acute lung injury
  • Improves survival rate in mice
  • Involved in metabolic enzyme/protease pathway
  • Involved in immunology/inflammation pathway
  • Involved in NF-κB pathway