Learn More
Apexbio Technology LLC Oxaloacetic acid 328-42-7 200mg
SDP
Catalog No. 5000090299
Encompass_Preferred
Oxaloacetic acid (CAS 328-42-7) is a small molecule metabolic intermediate involved in multiple biosynthetic pathways including the citric acid cycle It modulates mitochondrial metabolism and has been shown to reduce intracellular reactive oxygen species (ROS) levels thereby attenuating oxidative stress in cells Due to these properties oxaloacetic acid serves as a valuable tool in the investigation of metabolic disorders and mitochondrial dysfunction in biomedical research
Catalog No. 50-000-90299 Supplier Apexbio Technology LLC Supplier No. C7079200
Oxaloacetic acid (CAS 328-42-7) is a small molecule metabolic intermediate involved in multiple biosynthetic pathways including the citric acid cycle It modulates mitochondrial metabolism and has been shown to reduce intracellular reactive oxygen species (ROS) levels thereby attenuating oxidative stress in cells Due to these properties oxaloacetic acid serves as a valuable tool in the investigation of metabolic disorders and mitochondrial dysfunction in biomedical research