Learn More
Sigma Aldrich Fine Chemicals Biosciences Oxaloacetic acid powder, BioReagent, suitable for cell culture, suitable for insect cell culture, >=97% (HPLC) | 328-42-7 | MFCD00002592 | 100G
Catalog No. 501856161
Encompass
Oxaloacetic acid powder, BioReagent, suitable for cell culture, suitable for insect cell culture, >=97% (HPLC) | Purity: >=97% (HPLC) | Mol Wt: 132.07 | 328-42-7 | MFCD00002592 | 100G
Catalog No. 50-185-6161 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. O7753100G
Add to Cart
Add to Cart
Oxaloacetic acid powder, BioReagent, suitable for cell culture, suitable for insect cell culture, >=97% (HPLC) | Purity: >=97% (HPLC) | Mol Wt: 132.07 | 328-42-7 | MFCD00002592 | 100G