Learn More
Cayman Chemical Oxalomalic Acid (sodium salt), CAS: 89304-26-7, 10 mg
SDP
Catalog No. 502955097
Encompass_Preferred
Formed by the condensation of oxaloacetate with glyoxylate; inhibits aconitase activity at 1 mM, which interferes with IRP1 binding and decreases glutamate secretion in lens and retinal pigment epithelial cells
Catalog No. 50-295-5097 Supplier Cayman Chemical Supplier No. 1352110 MG
Formed by the condensation of oxaloacetate with glyoxylate; inhibits aconitase activity at 1 mM, which interferes with IRP1 binding and decreases glutamate secretion in lens and retinal pigment epithelial cells