Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oxalyl Bromide

Catalog No. 11303425G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-303-425G 25 g
11-303-4100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-303-425G Supplier Sigma Aldrich Supplier No. 11303425G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Oxalyl Bromide

Specifications

CAS 15219-34-8
Melting Point -19°C
Boiling Point 16°C to 17°C (10 mmHg, lit.)
Molecular Formula C2Br2O2
Linear Formula BrCOCOBr
Refractive Index n20/D 1.522
MDL Number MFCD00000113
Quantity 25 g
Synonym Ethanedioyl dibromide; NSC 96957
Molecular Weight (g/mol) 215.83 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.