Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Oxalyl Bromide Solution

Catalog No. 32323350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-323-350ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 32-323-350ML Supplier Sigma Aldrich Supplier No. 32323350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in synthetic applications of carbon-substituted iminium saltsReactant for: Synthesis of mutasynthons added to cultures of A. pretiosum for mutasynthetic generation of ansamitocin derivativesOxalic acid formation from hydroxyl radical substitutionsCyclization to produce CRF1 receptor antagonistsPreparation, optical and electrochemical studies of thiophene end capped olig(2,3-alkylthieno[3,4-b]pyrazine)Asymmetrical synthesis of glycosyl chlorides and bromides

Specifications

CAS 75-16-1
Density 0.856 g/mL at 25°C
Molecular Formula CH3BrMg
Linear Formula CH3MgBr
MDL Number MFCD00000041
Quantity 100 mL
Synonym Ethanedioyl dibromide solution; NSC 96957
Molecular Weight (g/mol) 119.24 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.