Learn More
Medchemexpress LLC Oxalyldihydrazide | 996-98-5 | 98.0% | 118.09 | 100 G
SDP
Catalog No. 5000454787
Encompass_Preferred
Oxalyldihydrazide is a crosslinking agent used in proteomics research. It can capture carbonylated proteins on the surface of a microchip. It is for research use only and not sold to patients.
  • Crosslinking agent in proteomics research.
  • Can capture carbonylated proteins on the surface of a microchip.
Catalog No. 50-004-54787 Supplier Medchemexpress LLC Supplier No. HYW012887100G
Oxalyldihydrazide is a crosslinking agent used in proteomics research. It can capture carbonylated proteins on the surface of a microchip. It is for research use only and not sold to patients.
  • Crosslinking agent in proteomics research.
  • Can capture carbonylated proteins on the surface of a microchip.