Learn More
Medchemexpress LLC Oxalyldihydrazide | 996-98-5 | 98.0% | 118.09 | 25 G
SDP
Catalog No. 5000366937
Encompass_Preferred
Oxalyldihydrazide is a crosslinking agent used in proteomics research. It has the ability to capture carbonylated proteins on the surface of a microchip.
  • Crosslinking agent in proteomics research.
  • Captures carbonylated proteins on the surface of a microchip.
Catalog No. 50-003-66937 Supplier Medchemexpress LLC Supplier No. 25GHYW012887
Oxalyldihydrazide is a crosslinking agent used in proteomics research. It has the ability to capture carbonylated proteins on the surface of a microchip.
  • Crosslinking agent in proteomics research.
  • Captures carbonylated proteins on the surface of a microchip.