Learn More
Medchemexpress LLC Oxalyldihydrazide | 996-98-5 | 98.0% | 118.09 | 500 G
SDP
Supplier:  Medchemexpress LLC HYW012887500G
Encompass_Preferred
Oxalyldihydrazide is a crosslinking agent used in proteomics research that can capture carbonylated proteins on the surface of a microchip. It is for research use only and not sold to patients.
  • Crosslinking agent in proteomics research
  • Can capture carbonylated proteins on the surface of a microchip
Catalog No. 50-004-54897