Learn More
Medchemexpress LLC HY-102033 1mg Medchemexpress, Oxamflatin CAS:151720-43-3 Purity:>98%
SDP
Catalog No. 501967961
Encompass_Preferred
Medchemexpress, HY-102033 1mg Oxamflatin CAS:151720-43-3 Oxamflatin (Metacept-3) is a potent HDAC inhibitor with an IC50 of 15.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7961 Supplier Medchemexpress LLC Supplier No. HY1020331MG
Medchemexpress, HY-102033 1mg Oxamflatin CAS:151720-43-3 Oxamflatin (Metacept-3) is a potent HDAC inhibitor with an IC50 of 15.7 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.