Learn More
Apexbio Technology LLC Oxamic acid 471-47-6 100mg
SDP
Catalog No. 502240610
Encompass_Preferred
Oxamic acid is a small-molecule inhibitor targeting lactate dehydrogenase A (LDH-A) It is designed to inhibit LDH-A thereby disrupting the conversion of pyruvate to lactate during glycolysis and interfering with the glycolytic pathway Oxamic acid exerts its biological activity primarily through inhibition of LDH-A activity Based on these pharmacological properties oxamic acid holds research potential in studies of tumor metabolism investigation of metabolic vulnerabilities in cancer cells and screening of potential anti-cancer agents targeting glycolytic pathways
This item is not returnable. View return policy
Catalog No. 50-224-0610 Supplier Apexbio Technology LLC Supplier No. B7909100
Oxamic acid is a small-molecule inhibitor targeting lactate dehydrogenase A (LDH-A) It is designed to inhibit LDH-A thereby disrupting the conversion of pyruvate to lactate during glycolysis and interfering with the glycolytic pathway Oxamic acid exerts its biological activity primarily through inhibition of LDH-A activity Based on these pharmacological properties oxamic acid holds research potential in studies of tumor metabolism investigation of metabolic vulnerabilities in cancer cells and screening of potential anti-cancer agents targeting glycolytic pathways