Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Organic Chemistry Oxamic acid | 25G | 471-47-6 | MFCD00008006

Catalog No. O375025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred

Oxamic acid , 25G

About this item:

CAS #: 471-47-6
MDL #: MFCD00008006
Purity: 98%
Chemical Formula: C2H3NO3
Molecular Weight: 89.05
UNSPSC Code: 12352100

Catalog No. O375025G Supplier Sigma Organic Chemistry Supplier No. O375025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxamic acid , 25G

About this item:

CAS #: 471-47-6
MDL #: MFCD00008006
Purity: 98%
Chemical Formula: C2H3NO3
Molecular Weight: 89.05
UNSPSC Code: 12352100

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.