Learn More
Apexbio Technology LLC Oxamic acid 471-47-6 500mg
SDP
Catalog No. 5000088015
Encompass_Preferred
Oxamic acid is a small-molecule inhibitor targeting lactate dehydrogenase A (LDH-A) It is designed to inhibit LDH-A thereby disrupting the conversion of pyruvate to lactate during glycolysis and interfering with the glycolytic pathway Oxamic acid exerts its biological activity primarily through inhibition of LDH-A activity Based on these pharmacological properties oxamic acid holds research potential in studies of tumor metabolism investigation of metabolic vulnerabilities in cancer cells and screening of potential anti-cancer agents targeting glycolytic pathways
Catalog No. 50-000-88015 Supplier Apexbio Technology LLC Supplier No. B7909500
Oxamic acid is a small-molecule inhibitor targeting lactate dehydrogenase A (LDH-A) It is designed to inhibit LDH-A thereby disrupting the conversion of pyruvate to lactate during glycolysis and interfering with the glycolytic pathway Oxamic acid exerts its biological activity primarily through inhibition of LDH-A activity Based on these pharmacological properties oxamic acid holds research potential in studies of tumor metabolism investigation of metabolic vulnerabilities in cancer cells and screening of potential anti-cancer agents targeting glycolytic pathways