Learn More
Medchemexpress LLC Oxamic acid (sodium) | 565-73-1 | 99.7% | 111.03 | 100 MG
SDP
Catalog No. 5000367012
Encompass_Preferred
Oxamic acid (oxamate) sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor. Oxamic acid sodium salt shows anti-tumor activity, anti-proliferative activity against cancer cells, and can induce apoptosis. It is for research use only and not sold to patients.
  • Inhibits lactate dehydrogenase-A (LDH-A)
  • Shows anti-tumor activity
  • Exhibits anti-proliferative activity against cancer cells
  • Can induce apoptosis
  • Suppresses proliferation, migration, and invasion of A2780 and SKOV3 cells
  • Induces cell cycle arrest in the G2/M phase in CNE-1 and CNE-2 cells
  • Increases ROS levels in NPC cells
  • Improves efficacy of tumor inhibition in vivo when combined with irradiation treatment
Catalog No. 50-003-67012 Supplier Medchemexpress LLC Supplier No. HYW013032100MG
Oxamic acid (oxamate) sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor. Oxamic acid sodium salt shows anti-tumor activity, anti-proliferative activity against cancer cells, and can induce apoptosis. It is for research use only and not sold to patients.
  • Inhibits lactate dehydrogenase-A (LDH-A)
  • Shows anti-tumor activity
  • Exhibits anti-proliferative activity against cancer cells
  • Can induce apoptosis
  • Suppresses proliferation, migration, and invasion of A2780 and SKOV3 cells
  • Induces cell cycle arrest in the G2/M phase in CNE-1 and CNE-2 cells
  • Increases ROS levels in NPC cells
  • Improves efficacy of tumor inhibition in vivo when combined with irradiation treatment