Learn More
Medchemexpress LLC Oxamic acid sodium | 565-73-1 | 99.7% | 111.03 | 100 MG
SDP
Catalog No. 5000451394
Encompass_Preferred
Oxamic acid sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor. It exhibits anti-tumor and anti-proliferative activities against cancer cells and can induce apoptosis. This product is for research use only.
  • Inhibits lactate dehydrogenase-A (LDH-A) activity.
  • Demonstrates anti-tumor and anti-proliferative activities.
  • Capable of inducing apoptosis in cancer cells.
  • Suppresses proliferation, migration, and invasion in various cell lines.
  • Induces G2/M cell cycle arrest.
  • Increases reactive oxygen species (ROS) levels in cells.
Catalog No. 50-004-51394 Supplier Medchemexpress LLC Supplier No. HYW013032A100MG
Oxamic acid sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor. It exhibits anti-tumor and anti-proliferative activities against cancer cells and can induce apoptosis. This product is for research use only.
  • Inhibits lactate dehydrogenase-A (LDH-A) activity.
  • Demonstrates anti-tumor and anti-proliferative activities.
  • Capable of inducing apoptosis in cancer cells.
  • Suppresses proliferation, migration, and invasion in various cell lines.
  • Induces G2/M cell cycle arrest.
  • Increases reactive oxygen species (ROS) levels in cells.