Learn More
Medchemexpress LLC Oxamic acid sodium (sodium oxamate) | 565-73-1 | 99.7% | 111.03 | 1 G
SDP
Catalog No. 5000452173
Encompass_Preferred
Oxamic acid sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor that exhibits anti-tumor and anti-proliferative activity against cancer cells, and can induce apoptosis.
  • Inhibits proliferation, migration, and invasion of A2780 and SKOV3 cells.
  • Induces cell cycle arrest in G2/M phase.
  • Promotes apoptosis via caspase-3 activation and mitochondrial pathway.
  • Increases reactive oxygen species (ROS) levels in NPC cells.
  • Enhances tumor inhibition in vivo when combined with irradiation.
Catalog No. 50-004-52173 Supplier Medchemexpress LLC Supplier No. HYW013032A1G
Oxamic acid sodium salt is a lactate dehydrogenase-A (LDH-A) inhibitor that exhibits anti-tumor and anti-proliferative activity against cancer cells, and can induce apoptosis.
  • Inhibits proliferation, migration, and invasion of A2780 and SKOV3 cells.
  • Induces cell cycle arrest in G2/M phase.
  • Promotes apoptosis via caspase-3 activation and mitochondrial pathway.
  • Increases reactive oxygen species (ROS) levels in NPC cells.
  • Enhances tumor inhibition in vivo when combined with irradiation.