Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxamyl, 95%, Thermo Scientific™

Catalog No. AC456140010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC456140010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC456140010 Supplier Thermo Scientific Chemicals Supplier No. 456140010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 23135-22-0
Molecular Formula C7H13N3O3S
Molecular Weight (g/mol) 219.26
InChI Key KZAUOCCYDRDERY-UITAMQMPSA-N
IUPAC Name (Z)-[(dimethylcarbamoyl)(methylsulfanyl)methylidene]amino N-methylcarbamate
SMILES CNC(=O)O\N=C(/SC)C(=O)N(C)C

Specifications

Assay Percent Range 95%
Quantity 1 g
Formula Weight 219.26
Chemical Name or Material Oxamyl
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement Fatal if swallowed.

Fatal if inhaled.

Harmful in contact with skin.

Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Rinse mouth.

Wear protective gloves/protective clothing.

IF ON SKIN: Wash with plenty of soap and water.

Do not breathe dust/fume/gas/mist/vapors/spray.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Immediately call a POISON CENTER or doctor/physician.

EINECSNumber 245-445-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.