Learn More
Medchemexpress LLC Oxanilide (N,N'-diphenyloxalamide) | 620-81-5 | MFCD00041758 | 240.26 g/mol | C14H12N2O2 | 100 G
SDP
Catalog No. 5000282870
Encompass_Preferred
Oxanilide (N,N'-diphenyloxalamide) is a small-molecule chemical intermediate used in organic synthesis and medicinal chemistry research, notably for constructing quinoline and disulfide derivatives. It is supplied as a solid and is intended for laboratory research applications; consult product documentation for handling and purity details.
  • Chemical intermediate for quinoline and disulfide derivative synthesis.
  • Solid form suitable for storage and handling in research laboratories.
  • Molecular weight 240.26 g/mol and chemical formula C14H12N2O2.
  • Available in multiple pack sizes, including 100 g, for scalability of experiments.
  • Intended for research use only and not for human use.
Catalog No. 50-002-82870 Supplier Medchemexpress LLC Supplier No. HYW140663100G
Oxanilide (N,N'-diphenyloxalamide) is a small-molecule chemical intermediate used in organic synthesis and medicinal chemistry research, notably for constructing quinoline and disulfide derivatives. It is supplied as a solid and is intended for laboratory research applications; consult product documentation for handling and purity details.
  • Chemical intermediate for quinoline and disulfide derivative synthesis.
  • Solid form suitable for storage and handling in research laboratories.
  • Molecular weight 240.26 g/mol and chemical formula C14H12N2O2.
  • Available in multiple pack sizes, including 100 g, for scalability of experiments.
  • Intended for research use only and not for human use.