Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxanilide 98.0+%, TCI America™
SDP

Catalog No. O008725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O008725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O008725G Supplier TCI America Supplier No. O008725G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 620-81-5
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00041758
InChI Key FTWUXYZHDFCGSV-UHFFFAOYSA-N
Synonym oxanilide, oxanilid, oxalanilide, n,n'-diphenylethanediamide, ethanediamide, n,n'-diphenyl, n,n'-diphenyloxalamide, oxaldianilide, oxamide, n,n'-diphenyl, ethanediamide, n1,n2-diphenyl, oxanilide 6ci,7ci,8ci
PubChem CID 69288
IUPAC Name N,N'-diphenylethanediamide
SMILES O=C(NC1=CC=CC=C1)C(=O)NC1=CC=CC=C1

Specifications

Melting Point 251°C
Color White-Yellow
Quantity 25 g
Formula Weight 240.26
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Oxanilide
RTECSNumber KH8368000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.