Learn More
TARGETMOL CHEMICALS INC Oxaprozin 200MG
SDP
Catalog No. 502880692
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Oxaprozin (Oxaprozinum) is a Nonsteroidal Anti-inflammatory Drug. The mechanism of action of oxaprozin is as a Cyclooxygenase Inhibitor. The chemical classification of oxaprozin is Nonsteroidal Anti-inflammatory Compounds. Purity 99.02%
Catalog No. 50-288-0692 Supplier TARGETMOL CHEMICALS INC Supplier No. T0708200MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Oxaprozin (Oxaprozinum) is a Nonsteroidal Anti-inflammatory Drug. The mechanism of action of oxaprozin is as a Cyclooxygenase Inhibitor. The chemical classification of oxaprozin is Nonsteroidal Anti-inflammatory Compounds. Purity 99.02%