Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Oxaprozin

Catalog No. AC463530250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC463530250 25 g
AC463530050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC463530250 Supplier Thermo Scientific Chemicals Supplier No. 463530250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 21256-18-8 | C18H15NO3 | 293.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21256-18-8
Molecular Formula C18H15NO3
Molecular Weight (g/mol) 293.32
MDL Number MFCD00215977
InChI Key OFPXSFXSNFPTHF-UHFFFAOYSA-N
IUPAC Name 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
SMILES OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 161.0°C to 165.0°C
Quantity 25 g
Solubility Information Solubility aq. soln.: insoluble. Other solubilities: soluble in alcohols, very slightly soluble in ether
Formula Weight 293.32
Chemical Name or Material Oxaprozin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

EINECSNumber 244-296-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.