Learn More
Medchemexpress LLC Oxaprozin-d4 | 2070015-11-9 | 99.1% | 1 MG
SDP
Catalog No. 5000409228
Encompass_Preferred
Oxaprozin-d4 is the deuterium labeled Oxaprozin, functioning as a non-steroidal anti-inflammatory agent (NSAID).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-09228 Supplier Medchemexpress LLC Supplier No. HYB0808S1MG
Oxaprozin-d4 is the deuterium labeled Oxaprozin, functioning as a non-steroidal anti-inflammatory agent (NSAID).
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.