Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxazine 1 perchlorate | 24796-94-9 | MFCD00041971 | 97.0% | 423.89 g·mol⁻¹ | C20H26ClN3O5 | 10 MG
SDP

Catalog No. 5000246993
Encompass_Preferred

Oxazine 1 perchlorate is a symmetric cationic fluorescent dye that absorbs near 653 nm and emits at about 666 nm. It is used for red-region fluorescence labeling and detection in research applications and is supplied as a solid.

  • Excitation at 653 nm and emission at 666 nm.
  • Molecular formula C20H26ClN3O5; molecular weight 423.89 g·mol⁻¹.
  • Typical purity about 97%.
  • Available in small-mass packs suitable for research use.
  • For research use only; not for clinical use or sale to patients.

Catalog No. 50-002-46993 Supplier Medchemexpress LLC Supplier No. HY10190110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxazine 1 perchlorate is a symmetric cationic fluorescent dye that absorbs near 653 nm and emits at about 666 nm. It is used for red-region fluorescence labeling and detection in research applications and is supplied as a solid.

  • Excitation at 653 nm and emission at 666 nm.
  • Molecular formula C20H26ClN3O5; molecular weight 423.89 g·mol⁻¹.
  • Typical purity about 97%.
  • Available in small-mass packs suitable for research use.
  • For research use only; not for clinical use or sale to patients.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.