Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Oxazine 170 perchlorate | 62669-60-7 | MFCD00042010 | 98.8% | 431.87 g/mol | C21H22ClN3O5 | 10 MG
SDP

Catalog No. 5000297188
Encompass_Preferred

Oxazine 170 perchlorate is the perchlorate salt of Oxazine 170, a laser dye and fluorescent probe. It exhibits a broad excitation range (300-550 nm) and emits around 645 nm. Supplied as a solid with high purity, it is intended for research use in fluorescence labeling, spectroscopy, and laser dye applications.

  • Perchlorate salt, solid form.
  • High purity (~98.8%).
  • Molecular weight 431.87 g/mol.
  • CAS number 62669-60-7.
  • Broad excitation range 300-550 nm; emission ≈645 nm.
  • Available in small mg pack sizes for research use.
  • Store sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Catalog No. 50-002-97188 Supplier Medchemexpress LLC Supplier No. HYD170310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Oxazine 170 perchlorate is the perchlorate salt of Oxazine 170, a laser dye and fluorescent probe. It exhibits a broad excitation range (300-550 nm) and emits around 645 nm. Supplied as a solid with high purity, it is intended for research use in fluorescence labeling, spectroscopy, and laser dye applications.

  • Perchlorate salt, solid form.
  • High purity (~98.8%).
  • Molecular weight 431.87 g/mol.
  • CAS number 62669-60-7.
  • Broad excitation range 300-550 nm; emission ≈645 nm.
  • Available in small mg pack sizes for research use.
  • Store sealed, away from moisture and light; in solvent: -80°C (6 months), -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.