Learn More
Medchemexpress LLC Oxazole | 288-42-6 | 100.0% | 69.06 | 1 G
SDP
Catalog No. 5000404501
Encompass_Preferred
Oxazole is a biochemical reagent for life science related research, suitable as a biological material or organic compound. Its derivatives are building blocks for biochemicals and pharmaceuticals such as flopristin and darglitazone. It also finds use in industrial applications including dyes, fluorescent brightening agents, textile auxiliaries, and plastics. This product is a highly flammable liquid and vapour that causes serious eye damage.
  • Biochemical reagent for life science research
  • Building block for biochemicals and pharmaceuticals
  • Used in industrial applications like dyes and plastics
  • Highly flammable liquid and vapour
  • Causes serious eye damage
Catalog No. 50-004-04501 Supplier Medchemexpress LLC Supplier No. HY346311G
Oxazole is a biochemical reagent for life science related research, suitable as a biological material or organic compound. Its derivatives are building blocks for biochemicals and pharmaceuticals such as flopristin and darglitazone. It also finds use in industrial applications including dyes, fluorescent brightening agents, textile auxiliaries, and plastics. This product is a highly flammable liquid and vapour that causes serious eye damage.
  • Biochemical reagent for life science research
  • Building block for biochemicals and pharmaceuticals
  • Used in industrial applications like dyes and plastics
  • Highly flammable liquid and vapour
  • Causes serious eye damage