Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oxazole-4-carboxylic acid, 97%

Catalog No. AC430110050
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC430110050 5 g
AC430110010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430110050 Supplier Thermo Scientific Chemicals Supplier No. 430110050
Add to Cart

CAS: 23012-13-7 | C4H3NO3 | 113.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23012-13-7
Molecular Formula C4H3NO3
Molecular Weight (g/mol) 113.07
MDL Number MFCD06797151
InChI Key JBCFJMYPJJWIRG-UHFFFAOYSA-N
Synonym oxazole-4-carboxylic acid, 4-oxazolecarboxylic acid, 4-oxazolecarboxylicacid, 4-carboxy-1,3-oxazole, oxazole-4-carboxylic, 4-oxazolecarboxlic acid, pubchem11025, 4-carboxyoxazole, 4-oxazolecarboxylic acid ep
PubChem CID 14281430
IUPAC Name 1,3-oxazole-4-carboxylic acid
SMILES C1=C(N=CO1)C(=O)O

Specifications

Melting Point 141.0°C to 144.0°C
Color Brown to Orange
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 113.07
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Oxazole-4-carboxylic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.