Learn More
Biotium OXAZOLE RED YO-PRO -3 1 MM
Catalog No. NC2373537
Encompass_Preferred
Oxazole Red (YO-PRO-3), 1 mM in DMSO
Catalog No. NC2373537 Supplier Biotium Supplier No. 40105
Oxazole Red (YO-PRO-3), 1 mM in DMSO