Learn More
NEOGEN Oxbile (Oxgall), 500 g
SDP
Catalog No. 502015798
Encompass_Preferred
Oxbile (Oxgall) is dehydrated bile for use in preparing microbiological culture media in a laboratory setting. Oxbile (Oxgall) is not intended for use in the diagnosis of disease or other conditions in humans. Oxbile is manufactured from large quantities of fresh bile by rapid evaporation of the water content. Bile is composed of fatty acids, bile acids, inorganic salts, sulfates, bile pigments, cholesterol, mucin, lecithin, glycuronicacids, porphyrins, and urea. The use of Oxbile insures a regular supply of bile, and uniformity impossible to obtain with fresh materials. Oxbile is dehydrated fresh bile and prepared specifically for differentiation of bile tolerant microorganisms. A 10% solution of dehydrated bile is equivalent to a fresh bile solution. It is usually incorporated into media e.g., Bile Esculin Agar and Brilliant Green Bile Agar, used for the determination of enteric pathogens.For more information visit https://www.neogen.com/solutions/Microbiology/oxbile-oxgall
Catalog No. 50-201-5798 Supplier NEOGEN Supplier No. 700003568
Oxbile (Oxgall) is dehydrated bile for use in preparing microbiological culture media in a laboratory setting. Oxbile (Oxgall) is not intended for use in the diagnosis of disease or other conditions in humans. Oxbile is manufactured from large quantities of fresh bile by rapid evaporation of the water content. Bile is composed of fatty acids, bile acids, inorganic salts, sulfates, bile pigments, cholesterol, mucin, lecithin, glycuronicacids, porphyrins, and urea. The use of Oxbile insures a regular supply of bile, and uniformity impossible to obtain with fresh materials. Oxbile is dehydrated fresh bile and prepared specifically for differentiation of bile tolerant microorganisms. A 10% solution of dehydrated bile is equivalent to a fresh bile solution. It is usually incorporated into media e.g., Bile Esculin Agar and Brilliant Green Bile Agar, used for the determination of enteric pathogens.For more information visit https://www.neogen.com/solutions/Microbiology/oxbile-oxgall